Learn More
Methyl chloroformate, 99%
CAS: 79-22-1 | C2H3ClO2 | 94.49 g/mol
$97.54 - $97.54
Chemical Identifiers
CAS | 79-22-1 |
---|---|
Molecular Formula | C2H3ClO2 |
Molecular Weight (g/mol) | 94.49 |
MDL Number | MFCD00000639 |
InChI Key | XMJHPCRAQCTCFT-UHFFFAOYSA-N |
Synonym | methyl chloroformate, methyl chlorocarbonate, chloroformic acid methyl ester, methoxycarbonyl chloride, carbonochloridic acid, methyl ester, methyl chloridocarbonate, chloro methoxy methanone, metilcloroformiato, k-stoff, methylchloorformiaat |
PubChem CID | 6586 |
IUPAC Name | methyl carbonochloridate |
SMILES | COC(Cl)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC126531000
|
Thermo Scientific Chemicals
126531000 |
100 mL | Glass bottle |
Each for $97.54
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentChemical Identifiers
79-22-1 | |
94.49 | |
XMJHPCRAQCTCFT-UHFFFAOYSA-N | |
6586 | |
COC(Cl)=O |
C2H3ClO2 | |
MFCD00000639 | |
methyl chloroformate, methyl chlorocarbonate, chloroformic acid methyl ester, methoxycarbonyl chloride, carbonochloridic acid, methyl ester, methyl chloridocarbonate, chloro methoxy methanone, metilcloroformiato, k-stoff, methylchloorformiaat | |
methyl carbonochloridate |
Specifications
79-22-1,75-44-5,67-56-1 | |
Colorless to Yellow | |
70.0°C to 72.0°C | |
Authentic | |
Glass bottle | |
1.3860 to 1.3880 | |
MFCD00000639 | |
03, 9 | |
1.223 | |
methyl chloroformate, methyl chlorocarbonate, chloroformic acid methyl ester, methoxycarbonyl chloride, carbonochloridic acid, methyl ester, methyl chloridocarbonate, chloro methoxy methanone, metilcloroformiato, k-stoff, methylchloorformiaat | |
XMJHPCRAQCTCFT-UHFFFAOYSA-N | |
methyl carbonochloridate | |
6586 | |
94.5 | |
Liquid |
-61.0°C | |
1.2230g/mL | |
5°C | |
98.5% min. (GC) | |
C2H3ClO2 | |
ClCO2CH3 | |
100 mL | |
06,376; 07,236; 09,306; 11,336 | |
15, 6114 | |
Solubility in water: hydrolysis. Other solubilities: miscible with alcohol, methanol, benzene,, chloroform and ether | |
COC(Cl)=O | |
94.49 | |
0.48 mPa.s (20°C) | |
99% | |
Methyl chloroformate, 97% |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Fatal if swallowed.
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Rinse mouth.
Immediately call a POISON CENT
GHS Signal Word: Danger
EINECSNumber : 201-187-3
Recommended Storage : Refrigerator +4°C
RUO – Research Use Only