missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-α-Methoxyphenylacetic acid, Honeywell Fluka™
for gravimetric determination of Na,≥99.0% (T)
$2,287.15 - $2,287.15
Chemical Identifiers
CAS | 7021-09-2 |
---|---|
Molecular Formula | C9H10O3 |
Molecular Weight (g/mol) | 166.18 |
MDL Number | MFCD00064217 |
InChI Key | DIWVBIXQCNRCFE-UHFFFAOYNA-N |
PubChem CID | 107202 |
IUPAC Name | 2-methoxy-2-phenylacetic acid |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7021-09-2 | |
166.18 | |
DIWVBIXQCNRCFE-UHFFFAOYNA-N | |
2-methoxy-2-phenylacetic acid |
C9H10O3 | |
MFCD00064217 | |
107202 |
Specifications
7021-09-2 | |
C9H10O3 | |
MFCD00064217 | |
NONH for all modes of transport | |
methoxyphenylacetic acid, methoxy phenyl acetic acid, dl-alpha-methoxyphenylacetic acid, acetic acid, methoxyphenyl, alpha-methoxyphenylacetic acid, o-methyl-dl-mandelic acid, benzeneacetic acid, .alpha.-methoxy, 2-methoxy-2-phenyl-acetic acid, methyloxy phenyl acetic acid, .alpha.-methoxyphenylacetic acid | |
COC(C1=CC=CC=C1)C(=O)O | |
166.18 | |
166.17g/mol | |
(±)-α-Methoxyphenylacetic acid |
Plastic Bottle | |
C6H5CH(OCH3)COOH | |
25 g | |
2209197 | |
DIWVBIXQCNRCFE-UHFFFAOYNA-N | |
2-methoxy-2-phenylacetic acid | |
107202 | |
≥99% (T) |
Safety and Handling
P261-P305 + P351 + P338