missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.9%, Honeywell Burdick & Jackson
$713.48 - $4260.43
Chemical Identifiers
CAS | 67-56-1 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
PubChem CID | 887 |
ChEBI | CHEBI:17790 |
IUPAC Name | methanol |
SMILES | CO |
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Specifications
67-56-1 | |
0.792 g/cm3 | |
19 L | |
Metal Can | |
MFCD00004595 | |
0.1 % max. | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04 | |
ACS Reagent | |
by GC Analysis |
100.0000% | |
64.7°C | |
11°C | |
CH4O | |
UN1230 | |
1098229 | |
Miscible in Water at 20°C | |
CO | |
32.04 | |
CHEBI:17790 | |
>99.9% | |
Methanol, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.9% |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol
EINECSNumber : 200-659-6