missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methanol CHROMASOLV™, gradient grade, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™
Pure-Pac™ II containers require the Micromatic MacroValve coupler for dispensing solvents, Z560723. Product filtered through a 0.2μm filter.
$220.73 - $1683.65
Chemical Identifiers
CAS | 67-56-1 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
PubChem CID | 887 |
ChEBI | CHEBI:17790 |
IUPAC Name | methanol |
SMILES | CO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
6000946
|
Honeywell-Chromasolv
348851L |
1 L |
Each for $220.73
|
|
|||||
6000947
|
Honeywell-Chromasolv
3488525L |
2.5 L |
Each for $246.23
|
|
|||||
6000951
|
Honeywell-Chromasolv
348856X1L |
6 x 1 L |
Case of 6 Each for $1,108.73
|
|
|||||
6000950
|
Honeywell-Chromasolv
348854X25L |
4 x 2.5 L |
Case of 4 Each for $790.78
|
|
|||||
6000959
|
Honeywell-Chromasolv
348854X4L |
4 x 4 L |
Case of 4 Each for $1,683.65
|
|
|||||
Description
- Same high quality as CHROMASOLV HPLC
- Additional testing for HPLC via gradient analysis and UV-VIS spectroscopy
- Low UV profile by HPLC
- Low impurity gradient LC baseline
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Specifications
-98°C | |
absorption/ in accordance | |
64.7°C | |
n20/D 1.329 | |
CH3OH | |
97.68mmHg (20°C) | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CO | |
32.04 | |
CHEBI:17790 | |
≥99.9% | |
Methanol, for HPLC |
67-56-1 | |
0.791g/mL at 25°C | |
1 L | |
CH4O | |
MFCD00004595 | |
1098229 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04g/mol | |
CHROMASOLV™ Gradient |
Safety and Handling
P210-P260-P280-P301 + P310-P311
H225-H301-H311-H331-H370
EINECSNumber : 200-659-6
RTECSNumber : PC1400000