missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metaphosphoric Acid, 3% w/v, Reagents
Metaphosphoric Acid, 3% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$762.73 - $11778.40
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | H2O |
Molecular Weight (g/mol) | 18.02 |
InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
Synonym | Phosphenic Acid |
IUPAC Name | water |
SMILES | O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-334-763
|
Labchem Inc
CM10350010F |
10 L |
Each for $7,852.23
|
|
|||||
01-334-762
|
Labchem Inc
CM10350020F |
20 L |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
01-334-761
|
Labchem Inc
CM1035004A |
4 L |
Each for $3,051.03
|
|
|||||
01-334-760
|
Labchem Inc
CM103500500A |
500 mL |
Each for $762.73
|
|
|||||
Chemical Identifiers
7732-18-5 | |
18.02 | |
Phosphenic Acid | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
Specifications
Colorless | |
100°C | |
1.019 | |
H2O | |
Phosphenic Acid | |
O | |
18.02 | |
Reagent |
7732-18-5,64-19-7 | |
Cubitainer | |
10 L | |
Liquid | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
3% | |
Metaphosphoric Acid, 3% w/v |