missing translation for 'onlineSavingsMsg'
Learn More

Mesityl Oxide, Approx. 98%, Spectrum™ Chemical

M2491, 141-79-7, C6H10O

$503.29 - $2886.83

Chemical Identifiers

CAS 141-79-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
InChI Key SHOJXDKTYKFBRD-UHFFFAOYSA-N
IUPAC Name 4-methylpent-3-en-2-one
SMILES CC(C)=CC(C)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18604334
View Documents
Spectrum Chemical Mfg Cor
M2491100ML
100 mL Amber Glass Bottle
Each for $503.29
Only null left
 
18604335
View Documents
Spectrum Chemical Mfg Cor
M2491500MLGL
500 mL Amber Glass Bottle
Each for $1,275.89
Only null left
 
18604336
View Documents
Spectrum Chemical Mfg Cor
M249125LTGL
2.5 L Glass Bottle
Each for $2,886.83
Only null left
 
Description

Description

Spectrum™ Chemical Mesityl Oxide is a colorless compound with a strong peppermint scent. It is known to be a volatile liquid. Mesityl oxide is produced under the same conditions of isophorone. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications

Chemical Identifiers

141-79-7
98.15
4-methylpent-3-en-2-one
C6H10O
SHOJXDKTYKFBRD-UHFFFAOYSA-N
CC(C)=CC(C)=O

Specifications

141-79-7
Amber Glass Bottle
100 mL
SHOJXDKTYKFBRD-UHFFFAOYSA-N
4-methylpent-3-en-2-one
∼98%
100%
C6H10O
UN1229
CC(C)=CC(C)=O
98.15
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'