missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Stretch Denim Flame Resistant Dungaree
Men's Stretch Denim Flame Resistant Dungaree fits like your weekend jeans and moves with you while you work. Relaxed through waist, hip, and thigh to allow for easy movement.
Supplier: Bulwark™ PSJ6SD4432
Description
- Increased durability with triple needle felled out seams and seat seam
- Sits below natural waist with relaxed fit through waist, hip, and thigh
- Bootcut leg opening fits easily over boots
- Five traditional jean style pockets, with an additional ruler pocket on right leg and utility pocket on left leg for extra storage
- One-piece waistband with metal button closure (metal is not exposed to skin)
- Utility hammer loop on left hip
- Reinforced pockets help prevent holes
- Hazard: Arc Flash, Flash Fire
- Number of Pockets: 7
- Primary Closure: Concealed Button
- Wash Care: Industrial Laundry - Light Soil
- Fabric: Body: 12 oz. (405 g/m2) Flame Resistant Denim
- Blend: Body: 94% Cotton/5% Polyester/1% Spandex Elastane
- Country of Origin: Imported, Made in USA Fabric
Specifications
44 x 32 in. | |
Cotton, Polyester, Spandex | |
Stitched | |
Concealed Button | |
Category 2 | |
NFPA 2112, NFPA 70E |
Men's | |
Denim | |
6 | |
Reusable | |
15 |