missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Relaxed Fit Bootcut Flame Resistant Jean with Stretch
Men's Relaxed Fit Bootcut Flame Resistant Jean with Stretch are made from 12 oz. flame-resistant 94% cotton, 5% polyester, 1% spandex denim that provides comfort and mobility while you work.
Supplier: Bulwark™ PSJ2SD3330
Description
- Sits below natural waist with relaxed fit through waist, hip and thigh
- Bootcut leg opening fits easily over boots
- One-piece waistband with metal button closure (metal is not exposed to skin)
- Two hip pockets and scoop front pockets
- Larger belt loop and waistband
- Contrasting, two-toned stitching
- Stretch fabric inseam and seat seam
- Hazard: Flash Fire, Arc Flash
- Wash Care: Industrial Laundry - Light Soil, Home Wash
- Fabric: Flame-resistant, 12 oz. (405 g/m2)
- Blend: 94% Cotton/5% Polyester/1% Spandex
- Country of Origin: Imported, Made in USA Fabric
Specifications
33 x 30 in. | |
Cotton, Polyester, Spandex | |
Stitched | |
Zipper | |
Category 2 | |
NFPA 2112 |
Men's | |
Denim | |
4 | |
Reusable | |
15 |