missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Long Sleeve Navy Flame Resistant Performance T-Shirt

Men's Long Sleeve Navy Flame Resistant Performance T-Shirt provides flame-resistant protection in a lightweight blended fabric. The result is unbeatable softness coupled with lasting durability. This casual FR shirt features raglan sleeves as well as rib knit collar and cuffs

$199.33 - $199.33

Specifications

Gender Men's
Material Modacrylic Blend
Color Navy
Neck Style Hemmed Crew
Sleeve Style Raglan Sleeve
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-204-255
View Documents
Bulwark™
SMT2NVRGM
Medium
Each for $199.33
Only null left
 
17-204-024
View Documents
Bulwark™
SMT2NVRGL
Large
Each for $199.33
Only null left
 
17-203-959
View Documents
Bulwark™
SMT2NVRGXL
X-Large
Each for $199.33
Only null left
 
17-204-263
View Documents
Bulwark™
SMT2NVRGXXL
2X-Large
Each for $199.33
Only null left
 
17-203-984
View Documents
Bulwark™
SMT2NVRG3XL
3X-Large
Each for $199.33
Only null left
 
Description

Description

  • As part of our CoolTouch™ 2 fabric collection, this long sleeve tee provides flame-resistant protection in a lightweight blended fabric.
  • The result is unbeatable softness coupled with lasting durability.
  • This casual FR shirt features raglan sleeves as well as rib knit collar and cuffs
  • Certifications Level 1: NFPA 2112, NFPA 70E
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Flash Fire, Arc Flash
  • Number of Pockets: 1
  • Primary Closure: No Closure
  • Wash Care: Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame-resistant, 6.5 oz. (220 g/m&superscript_2;) with moisture management finish
  • Blend: 45% Modacrylic / 35% FR Lyocell / 20% Aramid
  • Collar: Rib knit collar
  • Pocket: Single spade chest pocket
  • Features: Heat seal Bulwark™ triangle centered at neck to show wearer has on FR undergarment, Full side seam gusset, Rib knit cuffs and collar, Raglan sleeves for ease of movement, Tagless brand label and warning label
  • Product Collection: Cooltouch 2
  • Packaging: Hang tag
  • Protection: EBT Arc Rating: 8.2
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Regular Fit
  • Gender: Male
  • Materials: Modacrylic/FR Lyocell/Aramid
  • Brand: Bulwark™
Specifications

Specifications

Men's
Navy
Raglan Sleeve
Reusable
Category 2
NFPA 2112, NFPA 70E
Modacrylic Blend
Hemmed Crew
Pull On
1
8.2
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'