missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Lightweight Navy Flame Resistant T-Shirt
Men's Lightweight Navy Flame Resistant T-Shirt is a CAT 2 AR/FR, NFPA™ 2112 compliant tee that features a heat seal Bulwark™ triangle centered at neck to show wearer has on FR undergarment.
$99.74 - $99.74
Specifications
Gender | Men's |
---|---|
Material | FR Cotton |
Color | Navy |
Neck Style | Hemmed Crew |
Sleeve Style | Short Sleeve |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17-204-156
|
Bulwark™
SET8NVSSS |
Small |
Each for $99.74
|
|
|||||
17-204-036
|
Bulwark™
SET8NVSSM |
Medium |
Each for $99.74
|
|
|||||
17-204-152
|
Bulwark™
SET8NVSSL |
Large |
Each for $99.74
|
|
|||||
17-203-956
|
Bulwark™
SET8NVSSXL |
X-Large |
Each for $99.74
|
|
|||||
17-204-068
|
Bulwark™
SET8NVSSXXL |
2X-Large |
Each for $99.74
|
|
|||||
17-204-368
|
Bulwark™
SET8NVSS3XL |
3X-Large |
Each for $99.74
|
|
|||||
17-204-135
|
Bulwark™
SET8NVSS4XL |
4X-Large |
Each for $99.74
|
|
|||||
17-204-292
|
Bulwark™
SET8NVSSLL |
Large (Long) |
Each for $99.74
|
|
|||||
17-204-137
|
Bulwark™
SET8NVSSLXL |
X-Large (Long) |
Each for $99.74
|
|
|||||
17-204-069
|
Bulwark™
SET8NVSSLXXL |
2X-Large (Long) |
Each for $99.74
|
|
|||||
Description
- Meet the most comfortable FR short sleeve T-shirt you'll ever wear.
- This CAT 2 AR/FR, NFPA™ 2112 compliant tee features a heat seal Bulwark™ triangle centered at neck to show wearer has on FR undergarment.
- Certifications Level 1: ASTM F1506, NFPA 2112, NFPA 70E
- Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Flash Fire, Arc Flash
- Primary Closure: No Closure
- Wash Care: Home Wash
- Fabric: EXCEL FR™ Flame resistant, 7 oz. (235 g/m&superscript_2;) Interlock
- Blend: 100% Cotton
- Collar: Rib-knit crewneck collar
- Pocket: Single spade chest pocket
- Features: Hemmed sleeve, Heat seal Bulwark™ triangle centered at neck to show wearer has FR undergarment, Underarm gusset for improved breathability
- Product Collection: Excel FR
- Protection: ATPV Arc Rating: 8.9
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Regular Fit
- Gender: Male
- Materials: Cotton
- Brand: Bulwark™
Specifications
Men's | |
Navy | |
Short Sleeve | |
Reusable | |
Category 2 |
FR Cotton | |
Hemmed Crew | |
Pull On | |
1 | |
8.9 |