missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Lightweight Excel FR™ ComforTouch™ Navy Premium Flame Resistant Coverall

Men's Lightweight Excel FR™ ComforTouch™ Navy Premium Flame Resistant Coverall has eight total pockets to keep your tools at hand, while elastic waist inserts and deep-pleated action back provide added mobility and comfort while you work.

$218.10 - $218.10

Specifications

Product Type Flame-resistant Coveralls
Gender Men's
Material Cotton, Nylon
Color Navy
Disposable Reusable
View More Specs

Products 9
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Qty  
17-204-064
View Documents
Bulwark™
CLB2NVRGM
Medium
Each for $218.10
Only null left
 
17-204-376
View Documents
Bulwark™
CLB2NVRGL
Large
Each for $218.10
Only null left
 
17-204-167
View Documents
Bulwark™
CLB2NVRGXL
X-Large
Each for $218.10
Only null left
 
17-203-976
View Documents
Bulwark™
CLB2NVRGXXL
2X-Large
Each for $218.10
Only null left
 
17-204-470
View Documents
Bulwark™
CLB2NVRG3XL
3X-Large
Each for $218.10
Only null left
 
17-204-265
View Documents
Bulwark™
CLB2NVLNL
Large (Long)
Each for $218.10
Only null left
 
17-204-310
View Documents
Bulwark™
CLB2NVLNXL
X-Large (Long)
Each for $218.10
Only null left
 
17-204-315
View Documents
Bulwark™
CLB2NVLNXXL
2X-Large (Long)
Each for $218.10
Only null left
 
17-204-390
View Documents
Bulwark™
CLB2NVLN3XL
3X-Large (Long)
Each for $218.10
Only null left
 
Description

Description

  • This lightweight premium coverall provides CAT 2 FR/AR protection without added bulk.
  • Innovative features provide extra comfort and convenience.
  • Eight total pockets keep your tools at hand, while elastic waist inserts and deep-pleated action back provide added mobility and comfort while you work.
  • Certifications Level 2: Category 2 Protection, NFPA 2112 - 2018 Compliant
  • Hazard: Flash Fire, Arc Flash
  • Primary Closure: Concealed Zipper
  • Wash Care: Industrial Laundry - Heavy Soil, Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 7 oz. (235 g/m&superscript_2;)
  • Blend: 88% Cotton/ 12% Nylon
  • Closure: Two-way concealed Nomex™ taped brass break-away zipper, concealed snap at top of zipper and at neck
  • Pocket: Two front swing pockets are topstitched and lined and two patch hip pockets have single concealed snap closure, One left sleeve pocket, two patch chest pockets have flaps and concealed snap closure and a rule pocket on right leg
  • Features: Two-way concealed Nomex™ taped brass break-away zipper, concealed snap at top of zipper and at neck, Two-piece, lay-flay collar, deep-pleated action back and elastic waist inserts, Hemmed sleeve with adjustable snap and side vent opening with concealed snap closure
  • Product Collection: Excel FR Comfortouch
  • Protection: ATPV Arc Rating: 8.6
  • Country of Origin: Imported, Made in USA Fabric
  • Fit Details: Designed to be worn over other clothing. Fit over work boots.
  • Gender: Male
  • Materials: Cotton/Nylon
  • Brand: Bulwark™
Specifications

Specifications

Flame-resistant Coveralls
Cotton, Nylon
Reusable
Open
Collared
2-way Front Zipper with Storm Flap
Non-sterile
Flame
NFPA 2112
Men's
Navy
Elastic (Stitched-in)
Open
Unhooded
Stitched
CAT 2
8.6
8
Videos
SDS
Documents

Documents