missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark Men's FR/CP Lab Coat
Flame resistant/chemical protective lab coat brings together the latest innovations in lab safety to protect wearers against multiple hazards.
$319.75 - $319.75
Specifications
Gender | Men's |
---|---|
Material | Aramid Fiber |
Color | Royal Blue |
Closure Type | Snaps |
Cuff Style | Knit |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17-200-811
|
Bulwark
KNR6RBSM0R |
Small |
Each for $319.75
|
|
|||||
17-200-812
|
Bulwark
KNR6RBMD0R |
Medium |
Each for $319.75
|
|
|||||
17-200-813
|
Bulwark
KNR6RBLG0R |
Large |
Each for $319.75
|
|
|||||
17-200-814
|
Bulwark
KNR6RBXL0R |
X-Large |
Each for $319.75
|
|
|||||
17-200-815
|
Bulwark
KNR6RB2L0R |
2X-Large |
Each for $319.75
|
|
|||||
17-200-816
|
Bulwark
KNR6RB3L0R |
3X-Large |
Each for $319.75
|
|
|||||
17-200-817
|
Bulwark
KNR6RB4L0R |
4X-Large |
Each for $319.75
|
|
|||||
17-200-818
|
Bulwark
KNR6RB5L0R |
5X-Large |
Each for $319.75
|
|
|||||
Description
Flame resistant/chemical protective lab coat brings together the latest innovations in lab safety to protect wearers against multiple hazards.
- Lightweight and breathable fire resistant/chemical protective lab coat.
- Black lay-down collar for easy recognition as a CP lab coat.
- Splash resistant pass-through for inner pocket access.
- Flame-resistant 4.5 oz. Nomex IIIA fabric prevents clothing ignition during short-duration thermal incidents and emergency exposure to flame.
- Repels small amounts of liquid chemicals that are either dropped or splashed on the fabric, resisting penetration and wicking of an inadvertent, secondary liquid chemical splash.
- Concealed snap front neck closures.
- One double-stitched chest pocket and two double-stitched lower pockets with flaps
- Arc Rating ATPV 5.6 cal/cm2
- Category 1 Protection
- NFPA 2112 Compliant
Specifications
Men's | |
Royal Blue | |
Knit | |
Non-sterile | |
Regular | |
Lay-down |
Aramid Fiber | |
Snaps | |
Reusable | |
Flame, Chemicals | |
3 | |
Category 1, NFPA 2112 |