missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melatonin, Powder, 96%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor M2095100GM
Specifications
73-31-4 | |
Amber Glass Bottle | |
0.96 |
1 | |
100 g | |
Reagent |
Chemical Identifiers
73-31-4 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.