Learn More
Thermo Scientific Chemicals Megestrol acetate
$143.91 - $442.51
Chemical Identifiers
CAS | 595-33-5 |
---|---|
Molecular Weight (g/mol) | 384.52 |
InChI Key | RQZAXGRLVPAYTJ-GQFGMJRRSA-N |
IUPAC Name | (1R,3aS,3bR,9aR,9bS,11aS)-1-acetyl-5,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl acetate |
SMILES | CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(C)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC465790010
|
Thermo Scientific Chemicals
465790010 |
1 g |
Each for $143.91
|
|
|||||
AC465790050
|
Thermo Scientific Chemicals
465790050 |
5 g |
Each for $442.51
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
595-33-5 | |
RQZAXGRLVPAYTJ-GQFGMJRRSA-N | |
CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(C)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)=O |
384.52 | |
(1R,3aS,3bR,9aR,9bS,11aS)-1-acetyl-5,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl acetate |
Specifications
595-33-5 | |
C24H3204 | |
Solubility aq. soln.: .. Other solubilities: soluble in chloroform | |
Megestrol acetate |
210.0°C to 212.0°C | |
1 g | |
384.51 |
Safety and Handling
GHS H Statement Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 209-864-5
RUO – Research Use Only