missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Media Lab Bottle, Non-Graduated, Rubber Cap/Liner
Clear Type I borosilicate glass bottle without graduations.
$628.00 - $985.18
Specifications
Color | Clear |
---|---|
Material | Borosilicate Glass (Type I) |
Closure Color | Black |
Product Type | Media Bottle |
Closure Liner | Rubber |
Catalog Number | Mfr. No. | Capacity (Metric) | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Quantity | Price | Quantity | ||||
06-414
|
DWK Life Sciences
219575 |
125 mL | 48 EA |
Case of 48 Each for $628.00
|
|
||||
06-415
|
DWK Life Sciences
219577 |
250 mL | 48 EA |
Case of 48 Each for $985.18
|
|
||||
22-263-008
|
DWK Life Sciences
219579 |
500 mL | 24 EA |
Case of 24 Each for $637.32
|
|
||||
Description
- Manufactured from borosilicate glass which provides the best chemical resistance
- Clear borosilicate glass bottle
- Special pour lip facilitates pouring
- Packaged in partitioned carton to help prevent scratch marks on bottles
- Black Phenolic Rubber Lined Cap
- Hazardous communication label included with each case of bottles allows for content ID and safety notification
- Tamper evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse
Specifications
Clear | |
Black | |
Rubber |
Borosilicate Glass (Type I) | |
Media Bottle | |
Phenolic |