missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MCP-2 ProQuantum Immunoassay Kit
ELISA
$920.00 - $4150.00
Specifications
Accession Number | P80075 |
---|---|
Assay Range | 0.0128 to 5000 pg/mL |
Assay Sensitivity | <0.01 pg/mL |
Conjugate | FAM™ |
Detection Method | Taqman™ Real-time PCR |
Description
This antimicrobial gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of N-terminal cysteine residues of the mature peptide. This chemokine is a member of the CC subfamily which is characterized by two adjacent cysteine residues. This cytokine displays chemotactic activity for monocytes, lymphocytes, basophils and eosinophils. By recruiting leukocytes to sites of inflammation this cytokine may contribute to tumor-associated leukocyte infiltration and to the antiviral state against HIV infection.Specifications
P80075 | |
<0.01 pg/mL | |
Taqman™ Real-time PCR | |
ProQuantum | |
Human | |
qPCR Instruments | |
1 hr. | |
FAM™ | |
Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL | |
CCL8 (MCP2) | |
2 hrs |
0.0128 to 5000 pg/mL | |
FAM™ | |
Proximity-based Immunoassay | |
Plasma, Serum, Supernatant | |
Protein Quantitation | |
6355 | |
Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase | |
RUO | |
Frozen at -20°C | |
30 mins |