Learn More
3M™ Masking Tape, 2307
General purpose masking tape for holding, bundling, sealing, general paint masking and a vast number of other jobs where a pressure-sensitive tape is needed.
Supplier:  3M™ 7000123530
Catalog No. 29-636-4107
Only null left
Explore available promotions
Description
Description
  • 3M™ Masking Tape 2307 has a rubber adhesive for instant adhesion that sticks easily and helps reduce labour
  • It provides good holding power to help resist lifting or curling
  • The crepe paper provides conformability for easy handling and application
  • It is easily tearable yet resists slivering
  • In addition, it provides easy-to-use controlled unwind
  • This tape boasts good paint hold-out that resists bleed-through
Specifications
Specifications
Rubber
18 mm x 55 m
0.71 in.
Tan
60.14 yd.
18 mm
Product Suggestions
Product Suggestions