missing translation for 'onlineSavingsMsg'
Learn More

Manganese Chloride, Tetrahydrate, USP, 98-101%, Spectrum™ Chemical

M1106, 13446-34-9, MnCl2·4H2O

$511.31 - $68563.37

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18604183
View Documents
Spectrum Chemical Mfg Cor
M1106125GM
125 g Poly Bottle
Each for $511.31
Only null left
 
18604184
View Documents
Spectrum Chemical Mfg Cor
M1106500GM
500 g Poly Bottle
Each for $1,308.20
Only null left
 
18604185
View Documents
Spectrum Chemical Mfg Cor
M110625KG
2.5 kg Poly Bottle
Each for $3,393.74
Only null left
 
18604186
View Documents
Spectrum Chemical Mfg Cor
M110612KG
12 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18604187
View Documents
Spectrum Chemical Mfg Cor
M110650KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Manganese Chloride, Tetrahydrate, USP is indicated for use as a supplement to intravenous solutions given for total parenteral nutrition (TPN) to maintain Manganese serum levels. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

13446-34-9
197.90
CNFDGXZLMLFIJV-UHFFFAOYSA-L
O.O.O.O.[Cl-].[Cl-].[Mn++]
Cl2H8MnO4
MFCD00149792
manganese(2+) tetrahydrate dichloride

Specifications

13446-34-9
125 g
MFCD00149792
O.O.O.O.[Cl-].[Cl-].[Mn++]
197.90
USP
Poly Bottle
100%
Cl2H8MnO4
CNFDGXZLMLFIJV-UHFFFAOYSA-L
manganese(2+) tetrahydrate dichloride
98 to 101%
36.0-38.5%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'