missing translation for 'onlineSavingsMsg'
Learn More

Manganese Chloride Tetrahydrate (Crystalline/Certified ACS), Fisher Chemical™

$331.08 - $544.42

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
Synonym Manganese (II) Chloride
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
M87-100
SDS View Documents Promotion Details
Thermo Fisher Scientific
M87100
100 g Glass Bottle
Each for $331.08
Case of 6 Each for $1,986.48
Only null left
 
M87-500
SDS View Documents Promotion Details
Thermo Fisher Scientific
M87500
500 g Glass Bottle
Each for $544.42
Case of 6 Each for $3,266.52
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13446-34-9
197.90
CNFDGXZLMLFIJV-UHFFFAOYSA-L
manganese(2+) tetrahydrate dichloride
Cl2H8MnO4
MFCD00149792
Manganese (II) Chloride
O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

58°C
13446-34-9
Solid
98 to 101%
MFCD00149792
CNFDGXZLMLFIJV-UHFFFAOYSA-L
manganese(2+) tetrahydrate dichloride
197.91
Certified ACS
Glass Bottle
Manganese Chloride Tetrahydrate
Red
5.5
100 g
Cl2H8MnO4
Manganese (II) Chloride
O.O.O.O.[Cl-].[Cl-].[Mn++]
197.90
98 to 101%
Pass Test
2.01g/cm³
Videos
Safety and Handling

Safety and Handling

SDS
missing translation for 'documents'

missing translation for 'documents'