missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maleic Acid 99.0+%, TCI America™
$42.87 - $57.79
Chemical Identifiers
CAS | 110-16-7 |
---|---|
Molecular Formula | C4H4O4 |
Molecular Weight (g/mol) | 116.07 |
MDL Number | MFCD00063177 |
InChI Key | VZCYOOQTPOCHFL-OWOJBTEDSA-N |
Synonym | maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid |
PubChem CID | 444266 |
ChEBI | CHEBI:18300 |
IUPAC Name | (2E)-but-2-enedioic acid |
SMILES | OC(=O)\C=C\C(O)=O |
Chemical Identifiers
110-16-7 | |
116.07 | |
VZCYOOQTPOCHFL-OWOJBTEDSA-N | |
444266 | |
(2E)-but-2-enedioic acid |
C4H4O4 | |
MFCD00063177 | |
maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid | |
CHEBI:18300 | |
OC(=O)\C=C\C(O)=O |
Specifications
131°C | |
White | |
MFCD00063177 | |
maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid | |
OC(=O)\C=C\C(O)=O | |
116.07 | |
CHEBI:18300 | |
≥99.0% (T) | |
Crystalline Powder |
110-16-7 | |
C4H4O4 | |
3261 | |
VZCYOOQTPOCHFL-OWOJBTEDSA-N | |
(2E)-but-2-enedioic acid | |
444266 | |
116.07 | |
25 g | |
Maleic Acid |
Safety and Handling
EINECSNumber : (2)-1100
RTECSNumber : OM9625000
TSCA : Yes