missing translation for 'onlineSavingsMsg'
Learn More

Maleic Acid, NF, 99-101%, Spectrum™ Chemical

M1203, 110-16-7, C4H4O4

$176.00 - $18062.93

Chemical Identifiers

CAS 110-16-7
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00063177
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
IUPAC Name (2E)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(O)=O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18604146
View Documents
Spectrum Chemical Mfg Cor
M120325GM
25 g Amber Glass Bottle
Each for $176.00
Only null left
 
18604147
View Documents
Spectrum Chemical Mfg Cor
M1203100GM
100 g Amber Glass Bottle
Each for $367.40
Only null left
 
18604149
View Documents
Spectrum Chemical Mfg Cor
M1203500GM
500 g Amber Glass Bottle
Each for $995.90
Only null left
 
18604148
View Documents
Spectrum Chemical Mfg Cor
M120312KG
12 kg Fiber Drum
Each for $3,526.10
Only null left
 
18604145
View Documents
Spectrum Chemical Mfg Cor
M120350KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Maleic Acid, NF
Specifications

Chemical Identifiers

110-16-7
116.07
VZCYOOQTPOCHFL-OWOJBTEDSA-N
OC(=O)\C=C\C(O)=O
C4H4O4
MFCD00063177
(2E)-but-2-enedioic acid

Specifications

110-16-7
C4H4O4
UN3261
OC(=O)\C=C\C(O)=O
116.07
NF
0.1%
100%
MFCD00063177
VZCYOOQTPOCHFL-OWOJBTEDSA-N
(2E)-but-2-enedioic acid
99 to 101%
25 g
Amber Glass Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'