missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maleic Acid, (cis-Butenedioic Acid), Reagents
Maleic Acid, (cis-Butenedioic Acid), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$175.93 - $814.60
Chemical Identifiers
CAS | 110-16-7 |
---|---|
Molecular Formula | C4H4O4 |
Molecular Weight (g/mol) | 116.07 |
InChI Key | VZCYOOQTPOCHFL-OWOJBTEDSA-N |
Synonym | cis-Butenedioic Acid, Maleinic Acid, Toxilic Acid |
IUPAC Name | (2E)-but-2-enedioic acid |
SMILES | OC(=O)\C=C\C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-337-585
|
Labchem Inc
C2108500100C2 |
100 g |
Each for $175.93
|
|
|||||
01-337-584
|
Labchem Inc
C21085001KG |
1 kg |
Each for $814.60
|
|
|||||
Chemical Identifiers
110-16-7 | |
116.07 | |
cis-Butenedioic Acid, Maleinic Acid, Toxilic Acid | |
OC(=O)\C=C\C(O)=O |
C4H4O4 | |
VZCYOOQTPOCHFL-OWOJBTEDSA-N | |
(2E)-but-2-enedioic acid |
Specifications
130°C to 130.5°C | |
275°C | |
C4H4O4 | |
VZCYOOQTPOCHFL-OWOJBTEDSA-N | |
(2E)-but-2-enedioic acid | |
Reagent | |
Glass Bottle | |
1.59 |
110-16-7 | |
Colorless/White | |
cis-Butenedioic Acid, Maleinic Acid, Toxilic Acid | |
OC(=O)\C=C\C(O)=O | |
116.07 | |
100 g | |
Solid | |
Maleic Acid, (cis-Butenedioic Acid) |