missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate, Reagent grade, Honeywell
Reagent Grade, anhydrous,≥97%
$910.70 - $910.70
Chemical Identifiers
CAS | 7487-88-9 |
---|---|
Molecular Formula | MgO4S |
Molecular Weight (g/mol) | 120.36 |
MDL Number | MFCD00011110 |
InChI Key | CSNNHWWHGAXBCP-UHFFFAOYSA-L |
Synonym | magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot |
PubChem CID | 24083 |
ChEBI | CHEBI:32599 |
SMILES | [Mg++].[O-]S([O-])(=O)=O |
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7487-88-9 | |
120.36 | |
CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
24083 | |
[Mg++].[O-]S([O-])(=O)=O |
MgO4S | |
MFCD00011110 | |
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
CHEBI:32599 |
Specifications
7487-88-9 | |
2.5 kg | |
MFCD00011110 | |
magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot | |
[Mg++].[O-]S([O-])(=O)=O | |
120.36 | |
CHEBI:32599 | |
≥97% | |
0.1mmHg (20°C) | |
Magnesium Sulfate |
Anhydrous | |
MgO4S | |
NONH for all modes of transport | |
CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
magnesium;sulfate | |
24083 | |
120.37g/mol | |
Reagent | |
Vapor Density: <0.01 (vs air) |
Safety and Handling
EINECSNumber : 231-298-2
RTECSNumber : OM4500000