missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Sulfate, Anhydrous, USP, 99-100.5%, Spectrum™ Chemical
M1354, 7487-88-9, MgSO4
Supplier: Spectrum Chemical Mfg Cor M135425KG
Description
Spectrum™ Chemical Magnesium Sulfate, Anhydrous, USP is the raw material for a common magnesium-based, pharmaceutical preparation called Epsom salts. The Epsom salt formulations are for both topical and oral indications. The topical preparation is a gel for soothing aches and pains while the internal formulation is commonly used as saline laxative. In it′s anhydrous form, magnesium sulfate is used as a drying agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities..Specifications
7487-88-9 | |
5.0-9.2 | |
MgO4S | |
CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
magnesium(2+) sulfate | |
99 to 100.5% | |
2% |
100% | |
2.5 kg | |
MFCD00011110 | |
[Mg++].[O-]S([O-])(=O)=O | |
120.36 | |
USP | |
Poly Pail |
Chemical Identifiers
7487-88-9 | |
120.36 | |
CSNNHWWHGAXBCP-UHFFFAOYSA-L | |
[Mg++].[O-]S([O-])(=O)=O |
MgO4S | |
MFCD00011110 | |
magnesium(2+) sulfate |