missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.95% (metals basis), Thermo Scientific Chemicals
$120.21 - $1488.15
Chemical Identifiers
CAS | 7439-95-4 |
---|---|
Molecular Formula | Mg |
Molecular Weight (g/mol) | 24.30 |
MDL Number | MFCD00085308 |
InChI Key | JLVVSXFLKOJNIY-UHFFFAOYSA-N |
Synonym | magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs |
PubChem CID | 5462224 |
ChEBI | CHEBI:25107 |
IUPAC Name | magnesium |
SMILES | [Mg] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4329909
|
Thermo Scientific Chemicals
04329909 |
10 g |
Each for $120.21
|
|
|||||
AA4329918
|
Thermo Scientific Chemicals
04329918 |
50 g |
Each for $407.78
|
|
|||||
AA4329930
|
Thermo Scientific Chemicals
04329930 |
250 g |
Each for $1,488.15
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-95-4 | |
24.30 | |
JLVVSXFLKOJNIY-UHFFFAOYSA-N | |
5462224 | |
magnesium |
Mg | |
MFCD00085308 | |
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs | |
CHEBI:25107 | |
[Mg] |
Specifications
7439-95-4 | |
Odorless | |
Slug | |
MFCD00085308 | |
JLVVSXFLKOJNIY-UHFFFAOYSA-N | |
magnesium | |
5462224 | |
99.95% | |
(metals basis) |
10 g | |
≈0.7g/slug | |
Mg | |
magnesio, sheet, powdered, magnesio italian, turnings, rieke's active, ribbon, unii-i38zp9992a, hsdb 654, nitrate, acs | |
[Mg] | |
24.30 | |
CHEBI:25107 | |
Magnesium slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length |
Safety and Handling
P210-P223-P231+P232-P235+P410-P240-P241-P280-P335+P334-P370+P378s-P501c
H228-H252-H261
DOTInformation : Hazard Class: 4.1; Packaging Group: III
EINECSNumber : 231-104-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only