missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Perchlorate, Honeywell Fluka™
ACS Reagent
$788.48 - $788.48
Chemical Identifiers
CAS | 10034-81-8 |
---|---|
Molecular Formula | Cl2MgO8 |
Molecular Weight (g/mol) | 223.197 |
MDL Number | MFCD00011102 |
InChI Key | MPCRDALPQLDDFX-UHFFFAOYSA-L |
Synonym | anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french |
PubChem CID | 24840 |
IUPAC Name | magnesium;diperchlorate |
SMILES | [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
10034-81-8 | |
223.197 | |
MPCRDALPQLDDFX-UHFFFAOYSA-L | |
24840 | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2] |
Cl2MgO8 | |
MFCD00011102 | |
anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french | |
magnesium;diperchlorate |
Specifications
10034-81-8 | |
Cl2MgO8 | |
MFCD00011102 | |
anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french | |
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2] | |
223.197 | |
223.21g/mol | |
Magnesium Perchlorate |
500 g | |
Mg(ClO4)2 | |
1475 | |
MPCRDALPQLDDFX-UHFFFAOYSA-L | |
magnesium;diperchlorate | |
24840 | |
ACS Reagent |
Safety and Handling
P220-P261-P305 + P351 + P338
H272-H315-H319-H335
EINECSNumber : 233-108-3
RTECSNumber : SC8925000