missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva MabSelect™ SuRe 70 Protein A Resin
Protein A resin with high binding capacity for cost-efficient separation mAbs.
$895.77 - $4777.82
Specifications
Product Type | Affinity Resin |
---|---|
Ligand | Alkali-stabilized Protein A-derived |
Particle Size | 50 μm |
Catalog Number | Mfr. No. | Sample Volume | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Sample Volume | Price | Quantity | |||||
17-542-401
|
Cytiva
17542401 |
25 mL |
Each for $895.77
|
|
|||||
17-542-402
|
Cytiva
17542402 |
200 mL |
Each for $4,777.82
|
|
|||||
Description
MabSelect™ SuRe 70 protein A resin with high dynamic binding capacity (DBC), enables highly cost-efficient capture of monoclonal antibodies (mAbs). The resin uses the well-established SuRe protein A ligand with good alkaline stability providing stable processing of mAbs.
- Cost-efficient at low resin utilization, such as in clinical manufacturing.
- Highest DBC of any of our MabSelect SuRe™ resins
- Alkaline-stable, well-established protein A ligand, from the MabSelect SuRe resin, is stable when cleaned with 0.1 M NaOH for 200 cycles and approximately 100 cycles using 0.5 M NaOH.
- Excellent for use in short bed heights and rapid resin cycling mode to further improve productivity in protein A step.
DBC at different residence times
- The MabSelect™ SuRe 70 resin is based on a high-flow agarose base matrix with an average bead size of 50 μm.
- The optimized porosity enables a fast uptake of mAb, and the DBC is less dependent on residence time (RT) compared to other resins (e.g., MabSelect SuRe™ LX and MabSelect PrismA™ resins).
- The DBC for trastuzumab is higher than 70 g/L in a well-packed column at 6 min RT.
Alkaline-stable Protein A ligand
- As native protein A is sensitive to alkaline conditions, the ligand in MabSelect™ SuRe 70 resin, also used in the MabSelect SuRe™ resin, has been stabilized by engineering one of the IgG-binding domains of protein A.
- The MabSelect™ SuRe 70 resin is stable when cleaned with 0.1 M NaOH for 200 cycles and approximately 100 cycles using 0.5 M NaOH without performance loss.
Formats
- MabSelect™ SuRe 70 resin is available as bulk resin in 25 mL, 200 mL, 1 L, 5 L, and 10 L containers.
- The resin can be ordered in pre-packed column formats such as HiTrap™ columns (1 and 5 mL), HiScreen™ columns, and on request as production scale ReadyToProcess™ columns.
- These formats enable use from lab-scale to commercial-scale manufacturing.
Cytiva's mAb toolbox
- Process volumes, number of batches, target molecule, impurity profile, and cleaning protocols influence which chromatography resin will be most cost-efficient and productive in a given application.
- Cytiva can support you in a discussion about how to set up your process and to make your process more cost efficient.
- And with every Cytiva resin you choose, you have Cytiva quality, an extensive security of supply program, and expertise to go along with it.
Specifications
Affinity Resin | |
Alkali-stabilized Protein A-derived | |
50 μm |