missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Xylene 99.0+%, TCI America™
$55.92 - $82.02
Chemical Identifiers
CAS | 108-38-3 |
---|---|
Molecular Formula | C8H10 |
Molecular Weight (g/mol) | 106.17 |
MDL Number | MFCD00008536 |
InChI Key | IVSZLXZYQVIEFR-UHFFFAOYSA-N |
Synonym | m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150 |
PubChem CID | 7929 |
ChEBI | CHEBI:28488 |
IUPAC Name | 1,3-xylene |
SMILES | CC1=CC(C)=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
X001325ML
|
TCI America
X001325ML |
25 mL |
Each for $55.92
|
|
|||||
X0013500ML
|
TCI America
X0013500ML |
500 mL |
Each for $82.02
|
|
|||||
Chemical Identifiers
108-38-3 | |
106.17 | |
IVSZLXZYQVIEFR-UHFFFAOYSA-N | |
7929 | |
1,3-xylene |
C8H10 | |
MFCD00008536 | |
m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150 | |
CHEBI:28488 | |
CC1=CC(C)=CC=C1 |
Specifications
108-38-3 | |
139°C | |
Clear Liquid at 20°C | |
MFCD00008536 | |
m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150 | |
CC1=CC(C)=CC=C1 | |
106.17 | |
CHEBI:28488 | |
≥99.0% (GC) |
Colorless | |
25 mL | |
C8H10 | |
1307 | |
IVSZLXZYQVIEFR-UHFFFAOYSA-N | |
1,3-xylene | |
7929 | |
106.17 | |
m-Xylene |
Safety and Handling
EINECSNumber : (3)-0003&(3)-0060
RTECSNumber : ZE2275000
TSCA : Yes