missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Xylene, 98%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor X10154LTGL
Specifications
108-38-3 | |
Amber Glass Bottle | |
Reagent |
4 L | |
0.98 |
Chemical Identifiers
108-38-3 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.