Learn More
Applied Biosystems™ LV200™ dC(Bz) Pack
Contains 60 dC(Bz) ABI LV200™ synthesis columns.The dC(Bz) ABI LV200™ synthesis columns employ the advantages of polystyrene and a low volume column design to deliver high-quality oligonucleotides, while saving time and money.
Supplier: Applied Biosystems™ 4316441
Description
Save Time and MoneyCompared to conventional columns, the ABI LV200™ columns reduce cycle time by about 10% and the cost of synthesis by about 40%. These columns are fully compatible with the Applied Biosystems 3400 instruments, ABI 391, ABI 392, ABI 394, and earlier instruments.- Proprietary design and manufacturing methods ensure consistency and high quality
- Each column is tested to verify accuracy and precision of fill, then pressure-tested to verify proper assembly and to guarantee each column is leak-free
- Hydrophobic polystyrene columns sustain anhydrous environment to promote efficient coupling while minimizing nonspecific binding sites, yielding an efficient synthesis
- The 1,000 angstrom pore size of the polystyrene support allows synthesis of oligonucleotides of varying length
Specifications
Shipped at Room Temperature | |
60 Columns |
For Research Use Only. Not for use in diagnostic procedures.