missing translation for 'onlineSavingsMsg'
Learn More

Lumefantrine, 98%, Thermo Scientific™

Chemical Identifiers

CAS 82186-77-4
Molecular Formula C30H32Cl3NO
Molecular Weight (g/mol) 528.94
MDL Number MFCD05662268
InChI Key DYLGFOYVTXJFJP-MYYYXRDXNA-N
IUPAC Name 2-(dibutylamino)-1-[(9Z)-2,7-dichloro-9-[(4-chlorophenyl)methylidene]-9H-fluoren-4-yl]ethan-1-ol
SMILES CCCCN(CCCC)CC(O)C1=CC(Cl)=CC2=C1C1=CC=C(Cl)C=C1\C2=C\C1=CC=C(Cl)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC461110050
View Documents
Thermo Scientific Chemicals
461110050
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC461110250
View Documents
Thermo Scientific Chemicals
461110250
25 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

82186-77-4
528.94
DYLGFOYVTXJFJP-MYYYXRDXNA-N
CCCCN(CCCC)CC(O)C1=CC(Cl)=CC2=C1C1=CC=C(Cl)C=C1\C2=C\C1=CC=C(Cl)C=C1
C30H32Cl3NO
MFCD05662268
2-(dibutylamino)-1-[(9Z)-2,7-dichloro-9-[(4-chlorophenyl)methylidene]-9H-fluoren-4-yl]ethan-1-ol

Specifications

82186-77-4
C30H32Cl3NO
5 g
CCCCN(CCCC)CC(O)C1=CC(Cl)=CC2=C1C1=CC=C(Cl)C=C1\C2=C\C1=CC=C(Cl)C=C1
528.94
Lumefantrine
97.5 % min. (HPLC)
MFCD05662268
DYLGFOYVTXJFJP-MYYYXRDXNA-N
2-(dibutylamino)-1-[(9Z)-2,7-dichloro-9-[(4-chlorophenyl)methylidene]-9H-fluoren-4-yl]ethan-1-ol
528.94
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only