missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ LSE™ Shaking Incubator Accessory, Flat Platforms
For use with LSE 49L Shaking Incubator
$738.03 - $1460.88
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
07-202-169
|
Corning™
480154 |
Flat Platform |
Case for $738.03
|
|
|||||
07-202-170
|
Corning™
480155 |
Double Flat Platform |
Case for $1,460.88
|
|
|||||
Description
With nonslip rubber mat (300 x 300 mm)Specifications
LSE 49L Shaking Incubator |