missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Loratadine 98.0+%, TCI America™
$74.56 - $184.54
Chemical Identifiers
CAS | 79794-75-5 |
---|---|
Molecular Formula | C22H23ClN2O2 |
Molecular Weight (g/mol) | 382.888 |
MDL Number | MFCD00672869 |
InChI Key | JCCNYMKQOSZNPW-UHFFFAOYSA-N |
Synonym | loratadine, claritin, loratidine, alavert, clarityn, lisino, clarityne, loracert, loradex, bonalerg |
PubChem CID | 3957 |
IUPAC Name | ethyl 4-(8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate |
SMILES | CCOC(=O)N1CCC(=C2C3=C(CCC4=C2N=CC=C4)C=C(C=C3)Cl)CC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
L0223100MG
|
TCI America
L0223100MG |
100 mg |
Each for $74.56
|
|
|||||
L02231G
|
TCI America
L02231G |
1 g |
Each for $184.54
|
|
|||||
Chemical Identifiers
79794-75-5 | |
382.888 | |
JCCNYMKQOSZNPW-UHFFFAOYSA-N | |
3957 | |
CCOC(=O)N1CCC(=C2C3=C(CCC4=C2N=CC=C4)C=C(C=C3)Cl)CC1 |
C22H23ClN2O2 | |
MFCD00672869 | |
loratadine, claritin, loratidine, alavert, clarityn, lisino, clarityne, loracert, loradex, bonalerg | |
ethyl 4-(8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate |
Specifications
79794-75-5 | |
White | |
MFCD00672869 | |
loratadine, claritin, loratidine, alavert, clarityn, lisino, clarityne, loracert, loradex, bonalerg | |
CCOC(=O)N1CCC(=C2C3=C(CCC4=C2N=CC=C4)C=C(C=C3)Cl)CC1 | |
382.888 | |
382.89 | |
Crystalline Powder |
137°C | |
C22H23ClN2O2 | |
100 mg | |
JCCNYMKQOSZNPW-UHFFFAOYSA-N | |
ethyl 4-(8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate | |
3957 | |
≥98.0% (HPLC,T) | |
Loratadine |
Safety and Handling
RTECSNumber : TM6129200
TSCA : No