missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium hydroxide monohydrate, Honeywell Fluka™
ACS Reagent,≥98.0%
$287.55 - $287.55
Chemical Identifiers
CAS | 1310-66-3 |
---|---|
Molecular Formula | H3LiO2 |
Molecular Weight (g/mol) | 41.96 |
MDL Number | MFCD00149772 |
InChI Key | GLXDVVHUTZTUQK-UHFFFAOYSA-M |
Synonym | lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water |
PubChem CID | 168937 |
IUPAC Name | lithium;hydroxide;hydrate |
SMILES | [Li+].O.[OH-] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1310-66-3 | |
41.96 | |
GLXDVVHUTZTUQK-UHFFFAOYSA-M | |
168937 | |
[Li+].O.[OH-] |
H3LiO2 | |
MFCD00149772 | |
lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water | |
lithium;hydroxide;hydrate |
Specifications
1310-66-3 | |
H3LiO2 | |
MFCD00149772 | |
lithium hydroxide monohydrate, lithium hydroxide hydrate, lithium hydroxido, hydroxyde de lithium, unii-g51xlp968g, lithium hydroxide, monohydrate, lithiumhydrate, lioh-hydrate, lioh.hydrate, lioh water | |
[Li+].O.[OH-] | |
41.96 | |
41.96g/mol | |
Lithium hydroxide monohydrate |
500 g | |
LiOH · H2O | |
UN2680 | |
GLXDVVHUTZTUQK-UHFFFAOYSA-M | |
lithium;hydroxide;hydrate | |
168937 | |
≥98% |
Safety and Handling
P280-P305 + P351 + P338-P310