missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium chloride, Free-flowing, ACS reagent, anhydrous, ≥99%, Honeywell Fluka™
Free-flowing, ACS reagent, anhydrous,≥99%
$676.58 - $676.58
Chemical Identifiers
CAS | 7447-41-8 |
---|---|
Molecular Formula | ClLi |
Molecular Weight (g/mol) | 42.39 |
MDL Number | MFCD00011078 |
InChI Key | KWGKDLIKAYFUFQ-UHFFFAOYSA-M |
Synonym | lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride |
PubChem CID | 433294 |
ChEBI | CHEBI:48607 |
IUPAC Name | lithium;chloride |
SMILES | [Li+].[Cl-] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7447-41-8 | |
42.39 | |
KWGKDLIKAYFUFQ-UHFFFAOYSA-M | |
433294 | |
lithium;chloride |
ClLi | |
MFCD00011078 | |
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride | |
CHEBI:48607 | |
[Li+].[Cl-] |
Specifications
1,360°C | |
7447-41-8 | |
Anhydrous | |
ClLi | |
MFCD00011078 | |
KWGKDLIKAYFUFQ-UHFFFAOYSA-M | |
lithium;chloride | |
433294 | |
42.39 | |
ACS Reagent | |
2.07 g/cm3 |
605°C (literature) | |
≤100.0000% | |
500 g | |
LiCl | |
lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride | |
[Li+].[Cl-] | |
42.39 | |
CHEBI:48607 | |
>99% | |
Poly Bottle | |
Lithium chloride, Free-flowing, ACS reagent, anhydrous, ≥99% |
Safety and Handling
ShelfLife : 720 days from date of manufacture
EINECSNumber : 231-212-3
Recommended Storage : Room Temp