missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Light's Solution Oxidation-Reduction Potential (ORP) Standard, Reagents
Light's Solution Oxidation-Reduction Potential (ORP) Standard, Reagent, use for Redox. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CL000300500A
Specifications
Light's Solution Oxidation-Reduction Potential (ORP) Standard | |
7732-18-5,7783-85-9,7664-93-9 | |
Liquid | |
<1 | |
Completely miscible | |
Reagent |
Poly Bottle | |
1.0857 | |
Yellow | |
100°C | |
500 mL |