missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Tetrabutylammonium bisulfate,, For ion pair chromatography, ≥99.0%, MilliporeSigma™ Supelco™
Tetrabutyammonium bisulfate (TBAHS) is an ion-pairing reagent. It acts as a catalyst in hydrolysis of aziridines and epoxides and the catalyst can be recycled.
$296.64 - $1166.59
Chemical Identifiers
MDL Number | MFCD00011637 |
---|---|
Synonym | Tetrabutylammonium hydrogen sulfate |
Chemical Identifiers
MFCD00011637 | |
Tetrabutylammonium hydrogen sulfate |
Specifications
171°C to 174°C | |
175.6°C - Closed Cup | |
MFCD00011637 | |
Tetrabutylammonium hydrogen sulfate | |
≥99.0% (T); ≥99.0% | |
LiChropur |
32503-27-8 | |
(CH3 CH2 CH2 CH2)4 N(HSO4) | |
10 x 1 g | |
339.53 | |
For ion pair chromatography |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany