missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Sodium 1-decanesulfonate ,, For ion pair chromatography, ≥99.0%, MilliporeSigma™ Supelco™

$339.24 - $1313.63

Chemical Identifiers

CAS 13419-61-9
Molecular Formula C10H21NaO3S
Molecular Weight (g/mol) 244.32
MDL Number MFCD00007526
InChI Key AIMUHNZKNFEZSN-UHFFFAOYSA-M
Synonym 1-Decanesulfonic acid sodium salt
IUPAC Name sodium decane-1-sulfonate
SMILES [Na+].CCCCCCCCCCS([O-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111012972
View Documents
Merck Emd Millipore
3063110GF
10 g
Each for $339.24
Only null left
 
111012973
View Documents
Merck Emd Millipore
3063150GF
50 g
Each for $1,313.63
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13419-61-9
244.32
AIMUHNZKNFEZSN-UHFFFAOYSA-M
sodium decane-1-sulfonate
C10H21NaO3S
MFCD00007526
1-Decanesulfonic acid sodium salt
[Na+].CCCCCCCCCCS([O-])(=O)=O

Specifications

>300°C
10 g
MFCD00007526
244.33
Ion Pair Chromatography
13419-61-9
CH3(CH2)8 CH2 SO3 Na
1-Decanesulfonic acid sodium salt
≥99.0% (T); ≥99.0%
LiChropur
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany