missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, MilliporeSigma™ Supelco™

N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane is a silylating reagent generally used in estrogenic steroid hormone identification and quantification.

$110.18 - $813.96

Chemical Identifiers

MDL Number MFCD00008269
Synonym BSTFA + TMCS
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111019764
View Documents
Merck Emd Millipore
T63811AMP
1 x 1 mL
Each for $121.60
Only null left
 
111019960
View Documents
Merck Emd Millipore
T63815G
5 g
Each for $110.18
Only null left
 
111018754
View Documents
Merck Emd Millipore
T638110AMP
10 x 1 mL
Each for $284.96
Only null left
 
111018761
View Documents
Merck Emd Millipore
T638125G
25 g
Each for $284.96
Only null left
 
111018117
View Documents
Merck Emd Millipore
T6381100G
100 g
Each for $813.96
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00008269
BSTFA + TMCS

Specifications

25561-30-2
45°C to 50°C (14 mmHg)
Ampule
MFCD00008269
BSTFA + TMCS
For GC derivatization
0.97 g/mL
6°C
CF3 C[=NSi(CH3)3 ]OSi(CH3)3
1 x 1 mL
257.40
LiChropur
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany