missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ N-Methyl-N-(trimethylsilyl)trifluoroacetamide, BioReagent for Silylations, MilliporeSigma™ Supelco™
$163.50 - $539.27
Chemical Identifiers
MDL Number | MFCD00000411 |
---|---|
Synonym | N-Trimethylsilyl-N-methyl trifluoroacetamide; MSTFA |
Chemical Identifiers
MFCD00000411 | |
N-Trimethylsilyl-N-methyl trifluoroacetamide; MSTFA |
Specifications
24589-78-4 | |
130°C to 132°C | |
Ampule | |
MFCD00000411 | |
N-Trimethylsilyl-N-methyl trifluoroacetamide; MSTFA | |
199.25 | |
LiChropur |
1.07 g/mL at 25°C; 1.075 g/mL (at 25°C) | |
26°C | |
CF3 CON(CH3)Si(CH3)3 | |
5 g | |
>1 (vs. air) | |
8.8 mmHg (27°C) |
Safety and Handling
RTECSNumber : AC9800000
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany