missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ 1,1,1,3,3,3-Hexafluoro-2-Propanol, MilliporeSigma™ Supelco™
$392.92 - $1285.36
Chemical Identifiers
CAS | 920-66-1 |
---|---|
Molecular Formula | C3H2F6O |
Molecular Weight (g/mol) | 168.04 |
MDL Number | MFCD00011651 |
InChI Key | BYEAHWXPCBROCE-UHFFFAOYSA-N |
Synonym | HFP; Hexafluoroisopropanol |
IUPAC Name | 1,1,1,3,3,3-hexafluoropropan-2-ol |
SMILES | OC(C(F)(F)F)C(F)(F)F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
111022432
|
Merck Emd Millipore
1812710ML |
10 mL |
Each for $392.92
|
|
|||||
111022433
|
Merck Emd Millipore
1812750ML |
50 mL |
Each for $1,285.36
|
|
|||||
Chemical Identifiers
920-66-1 | |
168.04 | |
BYEAHWXPCBROCE-UHFFFAOYSA-N | |
1,1,1,3,3,3-hexafluoropropan-2-ol |
C3H2F6O | |
MFCD00011651 | |
HFP; Hexafluoroisopropanol | |
OC(C(F)(F)F)C(F)(F)F |
Specifications
920-66-1 | |
1.596 g/mL (at 25°C (literature)) | |
Closed Cup (Boils Before Flash) | |
(CF3)2 CHOH | |
MFCD00011651 | |
HFP; Hexafluoroisopropanol | |
OC(C(F)(F)F)C(F)(F)F | |
168.04 | |
≥99.8% (GC) |
-4°C (literature) | |
59°C (literature) | |
C3H2F6O | |
n20/D 1.275 (literature) | |
10 mL | |
BYEAHWXPCBROCE-UHFFFAOYSA-N | |
1,1,1,3,3,3-hexafluoropropan-2-ol | |
168.04 |
Safety and Handling
H314 - H361fd - H373
RTECSNumber : UB6450000
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany