missing translation for 'onlineSavingsMsg'
Learn More

LGR5 Mouse anti-Human, Clone:2B5B9, Abnova™

Mouse monoclonal antibody raised against recombinant human LGR5.

Supplier:  Abnova Corporation MAB17138

Catalog No. 89-333-501


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

LGR5
Monoclonal
Unconjugated
ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
LGR5
Mouse
100 μg
Human
Antibody
IgG2b
ELISA, Flow Cytometry, Western Blot
2B5B9
leucine-rich repeat-containing G protein-coupled receptor 5
In PBS (0.05% sodium azide)
FEX, GPR49, GPR67, GRP49, HG38, MGC117008
Recombinant protein corresponding to amino acid 22-178 of human LGR5 from E. coli.
8549
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions