missing translation for 'onlineSavingsMsg'
Learn More

Levocarnitine, Free Base, USP, 97-103%, Spectrum™ Chemical

C1399, 541-15-1, C7H15NO3

$517.87 - $70033.00

Chemical Identifiers

CAS 541-15-1
Molecular Formula C7H15NO3
Molecular Weight (g/mol) 161.20
InChI Key PHIQHXFUZVPYII-UHFFFAOYNA-N
IUPAC Name 3-hydroxy-4-(trimethylazaniumyl)butanoate
SMILES C[N+](C)(C)CC(O)CC([O-])=O
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18603801
View Documents
Spectrum Chemical Mfg Cor
C139925GM
25 g Poly Bottle
Each for $517.87
Only null left
 
18603802
View Documents
Spectrum Chemical Mfg Cor
C1399100GM
100 g Poly Bottle
Each for $1,466.73
Only null left
 
18603803
View Documents
Spectrum Chemical Mfg Cor
C1399500GM
500 g Poly Bottle
Each for $4,569.53
Only null left
 
18603804
View Documents
Spectrum Chemical Mfg Cor
C13991KG
1 kg Poly Pail
Each for $6,445.83
Only null left
 
18603805
View Documents
Spectrum Chemical Mfg Cor
C13995KGBL
5 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18603806
View Documents
Spectrum Chemical Mfg Cor
C139925KGBL
25 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Levocarnitine, Free Base, USP is used to prevent and treat a lack of carnitine. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

541-15-1
161.20
3-hydroxy-4-(trimethylazaniumyl)butanoate
C7H15NO3
PHIQHXFUZVPYII-UHFFFAOYNA-N
C[N+](C)(C)CC(O)CC([O-])=O

Specifications

541-15-1
C7H15NO3
PHIQHXFUZVPYII-UHFFFAOYNA-N
3-hydroxy-4-(trimethylazaniumyl)butanoate
97 to 103%
0.5%
100%
25 g
C[N+](C)(C)CC(O)CC([O-])=O
161.20
USP
Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'