missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ LanthaScreen™ Tb-labeled Anti-Goat Antibody Kit
Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents to facilitate in-house assay development around multiple target classes and assay formats.
$2185.00 - $22410.00
Specifications
Content And Storage | Store in freezer (-5 to -30°C). |
---|---|
Conjugate | Lanthanide Chelates, Terbium (Tb) |
Product Line | LanthaScreen™ |
Reactivity | Goat |
Shipping Condition | Dry Ice |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PV3771
![]() |
Thermo Scientific™
PV3771 |
1 mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
PV3769
![]() |
Thermo Scientific™
PV3769 |
25 μg |
Each for $2,185.00
|
|
|||||
Description
- Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents to facilitate in-house assay development around multiple target classes and assay formats
- Tb-labeled secondary antibodies bind to their corresponding species IgG with low nanomolar apparent Kd's, and show negligible cross reactivity with IgG's from other species
- Each Tb-labeled anti-species antibody is provided with a corresponding fluorescein-labeled IgG as a positive control
- In a kinase assay utilizing the Tb-labeled antibody generic reagent, anti-phosphopeptide primary antibody is not labeled
- The detection step is performed by the addition of the Tb-labeled secondary antibody
- This approach can be applied to other target classes and assay formats
Specifications
Store in freezer (-5 to -30°C). | |
LanthaScreen™ | |
Dry Ice |
Lanthanide Chelates, Terbium (Tb) | |
Goat |
For Research Use Only. Not for use in diagnostic procedures.