Learn More
Thermo Scientific™ LanthaScreen™ Tb-labeled Anti-Goat Ab DFS Item
This product is ideally suited for use during assay development when evaluating multiple primary goat antibodies, or when it is not possible to directly terbium label a primary antibody.
Supplier:  Thermo Scientific™ PV3770
 View more versions of this product
Catalog No. PV3770
Only null left
Explore available promotions
Description
Description
  • A general reagent for use in assay development, this terbium labeled antibody will bind goat antibodies
  • Ideally suited for use during assay development when evaluating multiple primary goat antibodies
  • Also for use when it is not possible to to directly terbium label a primary antibody
Specifications
Specifications
Lanthanide Chelates, Terbium (Tb)
Goat
End Point
25 μg
Terbium-labeled anti-Goat antibody in HEPES buffered saline. Store at -20°C.
Dry Ice
340/490, 520
LanthaScreen™
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.