missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) oxide, For AAS, ≥99.9%, MilliporeSigma™ Supelco™

Lanthanum (III) oxide is an alkaline earth oxide, and is a diamagnetic, isostructural analog of neodymium oxide. It is also an efficient recyclable catalyst, and can be used in coupling various aryl halides with aromatic/alkyl thiols.

$175.92 - $583.91

Chemical Identifiers

MDL Number MFCD00011071
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111012930
View Documents
Merck Emd Millipore
28920550G
50 g
Each for $175.92
Only null left
 
111012929
View Documents
Merck Emd Millipore
289205250G
250 g
Each for $583.91
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD00011071

Specifications

1312-81-8
50 g
MFCD00011071
≥99.9% (trace metals basis); ≥99.9%
Poly Bottle
Powder
La2O3
325.81
AAS
6.51 g/mL (at 25°C)
Videos
Safety and Handling

Safety and Handling

RTECSNumber : OE5330000

SDS
missing translation for 'documents'

missing translation for 'documents'