missing translation for 'onlineSavingsMsg'
Learn More

Lactic Acid, 1% v/v, Reagents

Lactic Acid, 1% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Labchem Inc CL1013004A

 View more versions of this product

Catalog No. 01-334-845

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Lactic Acid, 1% v/v
100°C
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
1%
4 L
Liquid
7732-18-5,50-21-5
Colorless
2-Hydroxypropanoic Acid, 2-hydroxypropionic Acid
O
18.02
Reagent
Poly Bottle
1.0021

Chemical Identifiers

7732-18-5
18.02
2-Hydroxypropanoic Acid, 2-hydroxypropionic Acid
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions