missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Protector™ Evidence Drying Cabinets with Washdown
Spray nozzle allows the interior to be washed down for easy clean up after the cabinet has been properly disinfected
Supplier: Labconco™ 3400010
Description
Key Features
- Provides a secure ventilated area to dry evidence such as clothing and bedding
- HEPA and carbon filtered exhaust protects personnel from exposure to biohazards while eliminating nuisance odors
Additional Features
- 2 in. (5.1cm) OD drain with 7.9 gal./min. (30L/min.) drain pump and washdown spray nozzle with 6 ft. (1.8m) coiled hose for additional decontamination
- Type 304 brushed stainless steel interior, stainless steel rod and three stainless steel collapsible/removable wire mesh shelves, 8W x 19 in.D (20 x 48cm)
- Safety glass door with magnetic seal that accommodates a user-supplied padlock or optional tamper-evident tag
- Epoxy-coated steel exterior
- Storage area, 17.5W x 26.5D x 12 in.H (44.5 x 67.3 x 30.5cm) with door
- 2 in. (5.1cm) diameter locking casters
- Inlet and exhaust roughing prefilters with MERV 11 rating; exhaust HEPA filter, 99.99% efficient on particles 0.3μm; activated organic vapor carbon filter, 7.5 lb. (3.4kg) porous coconut shell media
- Built-in blower with solid state speed control
- Red filter system alarm light to alert operator to low airflow conditions
- 96 in. (2.4m) power cord and plug
Specifications
36 in. | |
67.31 cm | |
Washdown system with pump | |
36 in. | |
26.5 in. | |
450 lb. | |
110 V | |
91.44cm | |
60 Hz | |
30.48 cm | |
Evidence Drying Cabinet | |
19 in. | |
Yes | |
Domestic | |
8 in. | |
48.26 cm | |
Stainless Steel |
200.7 cm | |
12 in. | |
Washdown system with pump | |
204 kg | |
Safety glass door with magnetic seal | |
71.1 cm | |
3 A | |
Evidence drying lockable cabinet | |
36 in. | |
91.44 cm | |
28 in. | |
30.48 cm | |
79 in. | |
HEPA and Carbon filter | |
91.44 cm | |
20.3 cm |