missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eagle™ Lab Pack Open Head Poly Drum with Bung Holes, 55 gal.
Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Bung lid includes two holes (3/4 in. and 2 in. diameters) and is ideal for safe storage, as well as handling or transportation of hazardous materials.
$156.94 - $232.89
Specifications
Product Type | Drum |
---|---|
Closure Included | Yes |
Capacity (English) | 55 gal. |
Capacity (Metric) | 208.48 L |
For Use With (Application) | Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration |
Catalog Number | Mfr. No. | Color | Closure Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Closure Type | Price | Quantity | ||||
19338474
|
Eagle™
1656MBLKBG |
Black | Metal Lever-lock |
Each for $156.94
|
|
||||
19338473
|
Eagle™
1656MBBG |
Blue | Metal Lever-lock |
Each for $223.38
|
|
||||
19338472
|
Eagle™
1656MBG |
Yellow | Metal Lever-lock |
Each for $232.89
|
|
||||
19338471
|
Eagle™
1656BLKBG |
Black | Plastic Lever-lock |
Each for $204.35
|
|
||||
Description
- Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
- For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
- Bung lid includes two holes, one of each diameter: 0.75 in. and 2 in.
- If used with leaking materials, drums must be packed with sufficient sorbents
- No free liquids
Specifications
Drum | |
55 gal. | |
Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration | |
Open | |
91 cm | |
23.5 lb. |
Yes | |
208.48 L | |
HDPE | |
35.8 in. | |
0.75 in., 2 in. (Bung Holes) |