Learn More
Thermo Scientific Chemicals L-Tyrosine, Cell Culture Reagent
CAS: 60-18-4 | C9H11NO3 | 181.19 g/mol
$43.78 - $880.89
Chemical Identifiers
CAS | 60-18-4 |
---|---|
Molecular Formula | C9H11NO3 |
Molecular Weight (g/mol) | 181.19 |
MDL Number | MFCD00002606 |
InChI Key | OUYCCCASQSFEME-SVGMAFHSNA-N |
Synonym | l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l |
PubChem CID | 6057 |
ChEBI | CHEBI:17895 |
SMILES | N[C@@H](CC1=CC=C(O)C=C1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6351109
|
Thermo Scientific Chemicals
J6351109 |
10 g |
Each for $43.78
|
|
|||||
AAJ6351122
|
Thermo Scientific Chemicals
J6351122 |
100 g |
Each for $164.18
|
|
|||||
AAJ6351136
|
Thermo Scientific Chemicals
J6351136 |
500 g |
Each for $584.32
|
|
|||||
AAJ63511A1
|
Thermo Scientific Chemicals
J63511A1 |
1 kg |
Each for $880.89
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
60-18-4 | |
181.19 | |
OUYCCCASQSFEME-SVGMAFHSNA-N | |
6057 | |
N[C@@H](CC1=CC=C(O)C=C1)C(O)=O |
C9H11NO3 | |
MFCD00002606 | |
l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l | |
CHEBI:17895 |
Specifications
>300°C | |
Brown to White | |
MFCD00002606 | |
14,9839 | |
OUYCCCASQSFEME-SVGMAFHSNA-N | |
(2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid | |
10 g | |
CHEBI:17895 | |
181.19 | |
Powder |
60-18-4 | |
C9H11NO3 | |
392441 | |
l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l | |
N[C@@H](CC1=CC=C(O)C=C1)C(O)=O | |
181.19 | |
6057 | |
−11° (c=4 in 1N HCl) | |
Cell Culture Reagent | |
L-Tyrosine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 200-460-4
RTECSNumber : YP2275600
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only